missing translation for 'onlineSavingsMsg'
Learn More

Azoxybenzene, 98+%, Thermo Scientific Chemicals

Product Code. 11430481
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430481 10 g 10g
11440481 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430481 Supplier Thermo Scientific Alfa Aesar Supplier No. B20021.09
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 495-48-7 | Molecular Formula: C12H10N2O | Molecular Weight: 198.225 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 495-48-7
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.225
MDL Number MFCD00019925
InChI Key GAUZCKBSTZFWCT-UHFFFAOYSA-N
Synonym azoxybenzene, azoxybenzide, azoxybenzol, fenazox, fentoxan, azobenzene, oxide, diphenyldiazene 1-oxide, benzene, azoxydi, diazene, diphenyl-, 1-oxide, ordinary azoxybenzene
PubChem CID 10316
ChEBI CHEBI:51865
IUPAC Name oxido-phenyl-phenyliminoazanium
SMILES C1=CC=C(C=C1)N=[N+](C2=CC=CC=C2)[O-]

Specifications

Melting Point 33°C to 36°C
Density 1.16
Boiling Point 130°C (0.9 mmHg)
Quantity 10 g
Beilstein 743984
Merck Index 14,923
Formula Weight 198.23
Percent Purity ≥98%
Chemical Name or Material Azoxybenzene
compliance-icons
Product Identifier
  • Azoxybenzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
Supplemental information
  • MIXTURE LIST-Contain: Azoxybenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.