missing translation for 'onlineSavingsMsg'
Learn More

Synergy Healthcare™ Azo™ Surface Disinfectant Spray

Product Code. 15659795
Change view
Click to view available options
Volume (Metric):
500 mL
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
15659795 500 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15659795 Supplier Synergy Healthcare™ Supplier No. 81120

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Kills bacteria effectively in under a minute. Synergy Healthcare™ Azo™ Surface Disinfectant Spray is Effective against variety of micro-organisms and pathogens.

  • Ideal for use in microbiology, critical care, hospital wards, dentists and cleanrooms and on wide range of surfaces
  • Fast contact time from 15 seconds
  • Offers 24 hour active protection against a wide range of micro-organisms
  • Foil sealed Bottle
  • Quick-drying formula
  • Contains isopropyl alcohol
  • Effective against variety of micro-organisms including MRSA, and is proved to be 99.9999% effective against pathogens validated by EN 13727

Specifications

Product Type Disinfectant Spray
Certifications/Compliance EN1276 and EN13727, EN1650/EN13624, EN14348
Composition Contains isopropyl alcohol
Container Type Foil sealed Bottle
For Use With (Application) For Universal use
Volume (Metric) 500 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.