missing translation for 'onlineSavingsMsg'
Learn More

Azomycin, MedChemExpress

Product Code. 16499895
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16499895 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16499895 Supplier Medchem Express Supplier No. HYN0195

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Azomycin (2-Nitroimidazole) is an antibiotic which can be active against aerobic Gram-positive and Gram-negative bacteria.

Azomycin (2-Nitroimidazole) is an antibiotic which can be active against aerobic Gram-positive and Gram-negative bacteria.

Specifications

Chemical Name or Material Azomycin
Physical Form Solid
CAS 527-73-1
Color Yellow
Quantity 250 mg
Purity Grade Notes Research
Molecular Formula C3H3N3O2
For Use With (Application) COVID-19-immunoregulation
Synonym 2-Nitroimidazole
Solubility Information DMSO : 160 mg/mL (1415.05 mM; Need ultrasonic)
SMILES O=[N+]([O-])C1=NC=CN1
Molecular Weight (g/mol) 113.07
Formula Weight 113.07
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.