missing translation for 'onlineSavingsMsg'
Learn More

Azomethine H monosodium salt monohydrate, 95%

Product Code. 15317667 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15317667

Brand: Thermo Scientific Acros 457090250

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 304655-82-1 | C17H15NNaO9S2 | 464.415 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 304655-82-1
Molecular Formula C17H15NNaO9S2
Molecular Weight (g/mol) 464.415
InChI Key CYPHYAYBFNUBEV-TTWKNDKESA-N
PubChem CID 131877441
IUPAC Name 4-hydroxy-5-[[(E)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]naphthalene-2,7-disulfonic acid;sodium;hydrate
SMILES C1=CC(=CNC2=C3C(=CC(=C2)S(=O)(=O)O)C=C(C=C3O)S(=O)(=O)O)C(=O)C=C1.O.[Na]

Specifications

Infrared Spectrum Authentic
Formula Weight 463.41
Percent Purity 95%
Quantity 25 g
Chemical Name or Material Azomethine H monosodium salt monohydrate
compliance-icons
Product Identifier
  • Azomethine H monosodium salt monohydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : Azomethine H monosodium salt monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.