missing translation for 'onlineSavingsMsg'
Learn More

Azomethine-H monosodium salt hydrate

Product Code. 11484323
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484323 25 g 25g
11464323 1 g 1g
11474323 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484323 Supplier Thermo Scientific Alfa Aesar Supplier No. A10927.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 206752-32-1 | C17H12NNaO8S2 | 445.39 g/mol

Azomethine-H monosodium salt hydrate is a reagent used for the colorimetric determination of boron in soil, plants, composts, manure, water and nutrient solutions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azomethine-H monosodium salt hydrate is a reagent used for the colorimetric determination of boron in soil, plants, composts, manure, water and nutrient solutions.

Solubility
Soluble in water, ethanol and acetone.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 206752-32-1
Molecular Formula C17H12NNaO8S2
Molecular Weight (g/mol) 445.39
MDL Number MFCD00010498
InChI Key GRYBZNLNHVSHPY-UHFFFAOYSA-M
Synonym sodium 5-hydroxy-4-2-hydroxybenzylidene amino-7-sulfonaphthalene-2-sulfonate hydrate, sodium 5-hydroxy-4-e-2-hydroxyphenyl methylidene amino-7-sulfonaphthalene-2-sulfonate hydrate, azomethin-h monosodium salt hydrate, 4-hydroxy-5-e-2-hydroxyphenyl methylidene amino-7-sodiooxysulfonyl naphthalene-2-sulfonic acid hydrate, sodium 5-hydroxy-4-2-hydroxyphenyl methylidene amino-7-sulfonaphthalene-2-sulfonate hydrate
PubChem CID 122198137
SMILES [Na+].OC1=C2C(NC=C3C=CC=CC3=O)=CC(=CC2=CC(=C1)S([O-])(=O)=O)S(O)(=O)=O

Specifications

Melting Point >300°C
Quantity 25 g
Beilstein 4100175
Solubility Information Soluble in water,ethanol and acetone.
IUPAC Name sodium;4-hydroxy-5-[[(Z)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]-7-sulfonaphthalene-2-sulfonate;hydrate
Formula Weight 445.41 (Anhydrous)
Chemical Name or Material Azomethine-H monosodium salt hydrate
compliance-icons
Product Identifier
  • Azomethine-H monosodium salt hydrate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Azomethine-H monosodium salt hydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.