missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azomethine-H Monosodium (>90%), TRC
Click to view available options
Quantity:
1 g
Unit Size:
1g
Specifications
Specifications
| Chemical Name or Material | Azomethine-H Monosodium |
| CAS | 5941-07-1 |
| Molecular Formula | C17H12NNaO8S2 |
| Quantity | 1 g |
| Synonym | 4-Hydroxy-5-(salicylidene-amino)-2,7-naphthalenedisulfonic Acid Monosodium, 4-Hydroxy-5-[[(2-hydroxyphenyl)methylene]amino]-2,7-naphthalenedisulfonic Acid Sodium Salt (1:1), 4-Hydroxy-5-(salicylideneamino)-2,7-naphthalenedisulfonic Acid Monosodium Salt |
| InChI Formula | InChI=1S/C17H13NO8S2.Na/c19-15-4-2-1-3-10(15)9-18-14-7-12(27(21,22)23)5-11-6-13(28(24,25)26)8-16(20)17(11)14;/h1-9,19-20H,(H,21,22,23)(H,24,25,26);/q;+1/p-1 |
| SMILES | OC1=CC(S(=O)(O[Na])=O)=CC2=CC(S(=O)(O)=O)=CC(/N=C/C3=C(O)C=CC=C3)=C21 |
| IUPAC Name | sodium;5-hydroxy-4-[(2-hydroxyphenyl)methylideneamino]-7-sulfonaphthalene-2-sulfonate |
| Molecular Weight (g/mol) | 445.4 |
| Formula Weight | 444.9902 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction