missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azomethine-H hydrate, Thermo Scientific Chemicals
Click to view available options
Quantity:
2 g
10 g
25 g
Description
Azomethine-H hydrate is used as a highly sensitive and colorimetric reagent for testing and detection of boron. It is also used to detect microgram levels of boron in glass and steel samples. Further, it is involved in the detection of boron in plant samples by masking copper, iron and aluminum ions with ethylenediaminetetraacetic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Solubility Information | Soluble in acetone,ammonium hydroxide,n-butanol and water. |
IUPAC Name | 4-hydroxy-5-[(Z)-[(2-hydroxyphenyl)methylidene]amino]naphthalene-2,7-disulfonic acid |
Formula Weight | 423.40 (Anhydrous) |
Sensitivity | Light sensitive |
Quantity | 25 g |
Physical Form | Powder |
Chemical Name or Material | Azomethine-H, hydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'