missing translation for 'onlineSavingsMsg'
Learn More

Azomethine-H hydrate, Thermo Scientific Chemicals

Product Code. 11327228
Click to view available options
Quantity:
2 g
10 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Reagent for boron determination

Azomethine-H hydrate is used as a highly sensitive and colorimetric reagent for testing and detection of boron. It is also used to detect microgram levels of boron in glass and steel samples. Further, it is involved in the detection of boron in plant samples by masking copper, iron and aluminum ions with ethylenediaminetetraacetic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32266-60-7
Molecular Formula C17H13NO8S2
Molecular Weight (g/mol) 423.41
MDL Number MFCD00066536
InChI Key DUCCKQSNXPFEGT-NVMNQCDNSA-N
PubChem CID 5702781
SMILES OC1=CC=CC=C1\C=N/C1=C2C(O)=CC(=CC2=CC(=C1)S(O)(=O)=O)S(O)(=O)=O

Specifications

Solubility Information Soluble in acetone,ammonium hydroxide,n-butanol and water.
IUPAC Name 4-hydroxy-5-[[(Z)-(6-oxocyclohexa-2,4-dien-1-ylidene)methyl]amino]naphthalene-2,7-disulfonic acid
Formula Weight 423.40 (Anhydrous)
Sensitivity Light sensitive
Quantity 2 g
Physical Form Powder
Chemical Name or Material Azomethine-H hydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'