missing translation for 'onlineSavingsMsg'
Learn More

Azocarmine G, TRC

Product Code. 30334419
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30334419 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30334419 Supplier Toronto Research Chemicals Supplier No. TRCA932808500MG
 Restricted Item
Estimated Shipment: 08-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Azocarmine G
CAS 25641-18-3
Molecular Formula C28 H18 N3 O6 S2 . Na
Quantity 500 mg
Synonym Benzo[a]phenazinium, 7-phenylsulfo-5-[(4-sulfophenyl)amino]-, hydroxide, inner salt, monosodium salt, Benzo[a]phenazinium, 7-phenylsulfo-5-[(4-sulfophenyl)amino]-, inner salt, monosodium salt (9CI), C.I. Acid Red 101 (7CI), C.I. Acid Red 101, monosodium s
InChI Formula InChI=1S/C28H19N3O6S2.Na/c32-38(33,34)20-12-10-18(11-13-20)29-25-17-27-28(22-15-14-21(16-23(22)25)39(35,36)37)30-24-8-4-5-9-26(24)31(27)19-6-2-1-3-7-19;/h1-17H,(H2,32,33,34,35,36,37);/q;+1/p-1
SMILES [Na+].[O-]S(=O)(=O)c1ccc(Nc2cc3c(nc4ccccc4[n+]3c5ccccc5)c6ccc(cc26)S(=O)(=O)[O-])cc1
IUPAC Name sodium;7-phenyl-5-(4-sulfonatoanilino)benzo[a]phenazin-7-ium-3-sulfonate
Molecular Weight (g/mol) 579.579
Formula Weight 579.053
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.