missing translation for 'onlineSavingsMsg'
Learn More

Azobenzene, MedChemExpress

Product Code. 16448738
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448738 10 g 10g
16412379 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16448738 Supplier Medchem Express Supplier No. HYB2127
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Azobenzene can be used as an optical trigger for the design and synthesis of a large variety of photoresponsive systems.

Azobenzene can be used as an optical trigger for the design and synthesis of a large variety of photoresponsive systems.

Specifications

Chemical Name or Material Azobenzene
CAS 103-33-3
Color Orange
Purity Grade Notes Research
Molecular Formula C12H10N2
Quantity 10 g
Solubility Information DMSO : ≥ 150 mg/mL (823.18 mM)
SMILES C1(/N=N/C2=CC=CC=C2)=CC=CC=C1
Molecular Weight (g/mol) 182.22
Formula Weight 182.22
Percent Purity 98.0%
Grade Research
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.