missing translation for 'onlineSavingsMsg'
Learn More

Azobenzene, 98%, Thermo Scientific Chemicals

Catalog Number p-3777355
Click to view available options
Quantity:
50 g
250 g
Packaging:
Glass bottle
This item is not returnable. View return policy
103-33-3
C12H10N2
182.23
MFCD00003022
DMLAVOWQYNRWNQ-UHFFFAOYSA-N
azobenzene, diazene, diphenyl, diazobenzene, e-diphenyldiazene, azobisbenzene, azofume, 1,2-diphenyldiazene, azodibenzeneazofume, diphenyldiimide, azobenzide
2272
CHEBI:190358
diphenyldiazene
C1=CC=C(C=C1)N=NC1=CC=CC=C1
This item is not returnable. View return policy
103-33-3
C12H10N2
182.23
MFCD00003022
DMLAVOWQYNRWNQ-UHFFFAOYSA-N
azobenzene, diazene, diphenyl, diazobenzene, e-diphenyldiazene, azobisbenzene, azofume, 1,2-diphenyldiazene, azodibenzeneazofume, diphenyldiimide, azobenzide
2272
CHEBI:190358
diphenyldiazene
C1=CC=C(C=C1)N=NC1=CC=CC=C1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 66.0°C to 69.0°C
Color Orange
Boiling Point 293.0°C
Flash Point 100°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5N=NC6H5
Quantity 50 g
Beilstein 16, 8
Merck Index 15, 909
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol, ether, glacial acetic acid,, acetone and benzene
Formula Weight 182.23
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material Azobenzene
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'