missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Spring Loaded Clamp

Product Code. 11327794
Click to view available options
Type:
Burette Clamp (Double)
Burette Clamp (Single)
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11327794

Brand: Azlon™ CWH002

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

For use with burettes. Azlon™ Burette Clamps contain rubber grips to prevent slipping.

For use with burettes. Azlon™ Burette Clamps contain rubber grips to prevent slipping.

  • Spring loaded polypropylene clamps that are corrosion resistant
  • Allow clear view of graduations along the full length of the burette
  • Rubber grips prevent slipping
  • An interchangeable compression cone on the central boss provides a firm grip on rods 11mm to 14mm in diameter
TRUSTED_SUSTAINABILITY

Specifications

Type Burette Clamp (Single)
Material Polypropylene
Includes Rubber grip, interchangeable compression cone
For Use With (Equipment) Burette
Color White
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.