Learn More
Azlon™ Silicone Rubber Hand/Finger Protector
Product Code. p-8001866
Click to view available options
Size:
Finger
Medium
Small
Unit Size:
Each
Product Code. 12018841
Brand: Azlon PGR406
Only null left
Add to basket
Add to basket
Ideal for safe handling of hot beakers, flasks
  • Capable of withstanding temperatures up to 250°C
  • For handling cold objects down to -57°C
  • Contact surfaces are multi-studded for positive grip
TRUSTED_SUSTAINABILITY
Specifications
Material Silicone
Product Type Hand/Finger Protector
Size Finger
For Use With (Application) For Safe Handling Of Hot Beakers, Flasks etc