missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Retort

Product Code. 12003374
Click to view available options
Mounting:
Center
Edge
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12003374

Brand: Azlon SWP014

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Polyethylene coated metal rod screws into the base.

Polyethylene coated metal rod screws into the base.

  • Weighs 2.5kg ensuring stability
  • Corrosion resistant
  • Available with the hole in the centre or at the short edge
TRUSTED_SUSTAINABILITY

Specifications

Dimensions (L x W x H) 171 x 248 x 52 mm (Base), 12.5 x 760 mm (Rod)
Mounting Edge
Product Type Retort
Description Rod Position: Edge
Quantity 1/Pk.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.