Learn More
Azlon™ PVDF and Polyethylene Stopcock
Product Code. 10115424
Click to view available options
Connectors:
2-way
3-way
Material:
PVDF
Polyethylene
For Use With (Equipment):
6 - 7mm I.D. tube
6 to 7mm I.D. tube
8 - 9mm I.D tube
8 to 9mm I.D. tube
Unit Size:
Each
Product Code. 10115424
Brand: Azlon SWQ206
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Range of two and three way stopcocks on PVDF or polyethylene
  • PVDF has excellent temperature, chemical and ageing resistance
  • Polyethylene available for less stringent requirements
TRUSTED_SUSTAINABILITY
Specifications
Material Polyethylene
Autoclavable Not Autoclavable
No. per Pack 1
For Use With (Equipment) 6 to 7mm I.D. tube
Connectors 3-way