Learn More
Azlon™ PTFE Stirrer Bars
Ideal for use with large vessels.
Brand: Azlon SWN680
Additional Details : Weight : 0.87000kg
Description
- Similar to cylindrical stirrer bars but give greater turbulence at low speed
Specifications
27 mm | |
2 | |
Magnet | |
108 mm |