Learn More
Azlon™ PTFE Stirrer Bars
Ideal for use with large vessels.
Brand: Azlon SWN677
Additional Details : Weight : 0.00800kg
Description
- Similar to cylindrical stirrer bars but give greater turbulence at low speed
Specifications
13 mm | |
2 | |
Magnet | |
75 mm |