missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azlon™ Polypropylene Autoclave bags
Description
- Suitable for autoclaving biohazardous waste
- Strong side weld construction (50μm)
- Printed with instructions for use and biohazard symbol
Specifications
Specifications
| Color | Blue |
| Height (Metric) | 608 mm |
| Width (Metric) | 300 mm |
| Autoclavable | Autoclavable |
| Material | Polypropylene |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?