missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azlon™ Polycarbonate Hinged Safety Shield
Protect yourself from potential hazards with a light weight but very strong safety shield. The Azlon™ polycarbonate hinged safety shield can be secured to a lab bench.
Brand: Azlon SGT002
Code : 03
Additional Details : Weight : 2.45000kg
Description
- Excellent protection in potentially hazardous situations
- Adjustable - can be used as a tight triangle or extended out to a straight line configuration
- Incredibly strong and lightweight
- Supplied with two pairs of plastic coated metal feet
- Feet may be clamped or bolted onto the bench to give greater stability
Specifications
Hinged safety shield | |
38 cm | |
Clear | |
1EA |
Polycarbonate | |
590 mm | |
For excellent protection in potentially hazardous situations | |
1EA |