missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azlon™ Polycarbonate Hinged Safety Shield
Description
- Excellent protection in potentially hazardous situations
- Adjustable - can be used as a tight triangle or extended out to a straight line configuration
- Incredibly strong and lightweight
- Supplied with two pairs of plastic coated metal feet
- Feet may be clamped or bolted onto the bench to give greater stability
Specifications
Specifications
| Product Type | Hinged safety shield |
| Material | Polycarbonate |
| Length (Metric) | 61 cm |
| Width (Metric) | 590 mm |
| Color | Clear |
| For Use With (Application) | For excellent protection in potentially hazardous situations |
| Quantity | 1EA |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?