missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Pipet Controller

Product Code. 11930885
Change view
Click to view available options
Color:
Blue
White
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color Unit Size
11930885 Blue Each
11303714 White Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11930885 Supplier Azlon Supplier No. PFL001B
  • ONLINE EXCLUSIVE PRICE
    £283.00
    £100.34 / Pack of 1
    Save £182.66
    64% Off
View all promotions
In Stock
Add to basket
Add to basket
Bulk Quote Request

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Easy and precise vacuum controlled thumb level pipet filler. The Azlon™ Pipet Controller uses a novel PTFE and silicone rubber valve assembly to allow fine control of pipent filling and emptying.

Easy and precise vacuum controlled thumb level pipet filler. The Azlon™Pipet Controller uses a novel PTFE and silicone rubber valve assembly to allow fine control of pipent filling and emptying.

  • A novel PTFE and silicone rubber valve assembly controls the vacuum created by simply squeezing the unique rubber bulb
  • By moving the thumb lever pipettes from 0.1mL up to 100mL can be easily and accurately filled and emptied
  • Blow out pipets are catered for by the depression of a small thumb operated squeeze ball
  • A replaceable PTFE membrane filter ensures liquid is not accidentally drawn into the unit (2 supplied)
  • The filter and pipet holding cone can be sterilized by autoclaving

Specifications

Material PVDF, Stainless Steel
Sterility Sterile
For Use With (Application) Clinical, hospital, biological, chemical, food and beverage, pharmaceutical, cosmetics and forensics fields
Color Blue
Volume (Metric) 0.1 to 100 mL
Type Positive displacement repetitive pipet
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.