Learn More
Azlon™ LDPE Wash Bottle, Bleach
Product Code. 15582656
Click to view available options
Capacity (Metric):
500 mL
Unit Size:
Pack of 5
Product Code. 15582656
Brand: Azlon WGW401PML
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Manufactured from opaque LDPE with yellow polypropylene jet-tip aperture closure. Azlon™ LDPE Wash Bottle, Bleach are made to contain bleach, and should not be used to contain any other chemicals or liquids.
  • Multi-lingual wide neck wash bottle
  • Opaque white LDPE for light sensitive sodium hypochlorite / bleach
  • Indelibly printed in three languages - English, French and Spanish
  • Features the new Globally Harmonized Symbols, NFPA diamond, risk phrases and safety information
  • Fitted with colour coded one-piece yellow PP closure
  • Closure features precision jet-tip aperture for fine control
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 500 mL
Color Opaque White
Material LDPE
Quantity 1Each
Closure Color Yellow
Mouth Wide
Chemical Label Sodium Hypochlorite (Bleach)
Closure Material Polypropylene
Closure Type Screw Cap with Jet-tip Aperture