missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azlon™ LDPE DIY Printed Safety Wash Bottles
Click to view available options
Capacity (Metric):
250 mL
500 mL
1000 mL
Closure Color:
Assorted
White
Description
Create custom labels for these do-it-yourself LDPE wash bottles. Azlon™ LDPE DIY Printed Safety Wash Bottles come with adhesive hazard labels and are ideal for a variety of liquid storage and sampling applications. DripLok™ vapor venting caps provide increased safety with volatile solvents.
- Do-it-yourself wash bottles with blank write-on panel
- Indelibly printed in three languages (English, French and German)
- Supplied with adhesive hazard labels
Specifications
Closure Material | Polypropylene |
Closure Type | Vapor-venting |
Diameter (Metric) | 90 mm |
Height (Metric) | 202 mm |
Capacity (Metric) | 1000 mL |
Color | Translucent |
Diameter (Metric) Outer Neck | 38 mm |
Diameter (Metric) Inner Neck | 34 mm |
Includes | Closure, blank write-on panel, adhesive hazard signs |
Material | LDPE |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'