missing translation for 'onlineSavingsMsg'
Learn More

Azlon™ Bottle Wide Neck 250mL 37mm HDPE Bulk Pack

Product Code. 15367027
Click to view available options
Capacity (Metric):
250 mL
Unit Size:
Pack of 200
This item is not returnable. View return policy

Product Code. 15367027

Brand: Azlon BWH0250BULK

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

HDPE bottles suitable for food use applications

This product is suitable for food contact applications in accordance with EU and\or USA regulations
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 250 mL
Material HDPE
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.