missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azlocillin sodium salt, MedChemExpress
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
Azlocillin sodium salt (Sodium azlocillin), a semisynthetic penicillin, is a broad spectrum β-lactam antibiotic. Azlocillin sodium salt shows antipseudomonal activity, and also potent against the malarial parasite Plasmodium falciparum.
Specifications
Specifications
| Chemical Name or Material | Azlocillin sodium salt |
| Physical Form | Solid |
| CAS | 37091-65-9 |
| Color | White |
| Quantity | 5 g |
| Purity Grade Notes | Research |
| Molecular Formula | C20H22N5NaO6S |
| For Use With (Application) | COVID-19-immunoregulation |
| Synonym | Sodium azlocillin |
| Solubility Information | DMSO : 100 mg/mL (206.84 mM; Need ultrasonic) ∣H2O : 6.67 mg/mL (13.80 mM; Need ultrasonic) |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction