missing translation for 'onlineSavingsMsg'
Learn More

Azlocillin sodium salt, MedChemExpress

Product Code. 16452219
Encompass_Preferred
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 16452219

Brand: Medchem Express HYB0529A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Azlocillin sodium salt (Sodium azlocillin), a semisynthetic penicillin, is a broad spectrum β-lactam antibiotic. Azlocillin sodium salt shows antipseudomonal activity, and also potent against the malarial parasite Plasmodium falciparum.

Azlocillin sodium salt (Sodium azlocillin), a semisynthetic penicillin, is a broad spectrum β-lactam antibiotic. Azlocillin sodium salt shows antipseudomonal activity, and also potent against the malarial parasite Plasmodium falciparum.

Specifications

Chemical Name or Material Azlocillin sodium salt
Physical Form Solid
CAS 37091-65-9
Color White
Quantity 1 g
Purity Grade Notes Research
Molecular Formula C20H22N5NaO6S
For Use With (Application) COVID-19-immunoregulation
Synonym Sodium azlocillin
Solubility Information DMSO : 100 mg/mL (206.84 mM; Need ultrasonic) ∣H2O : 6.67 mg/mL (13.80 mM; Need ultrasonic)
SMILES O=C([C@@H](C(C)(C)S[C@]1([H])[C@@H]2NC([C@H](NC(N3CCNC3=O)=O)C4=CC=CC=C4)=O)N1C2=O)O[Na]
Molecular Weight (g/mol) 483.47
Formula Weight 483.47
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.