missing translation for 'onlineSavingsMsg'
Learn More

Azithromycin N-Ethyl, TRC

Product Code. 30135739
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135739 1 g 1g
30006347 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30135739 Supplier Toronto Research Chemicals Supplier No. TRCA9270101G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92594-45-1
Molecular Formula C39H74N2O12
Molecular Weight (g/mol) 763.01
Synonym [2R-(2R*,3S*,4R*,5R*,8R*,10R*,11R*,12S*,13S*,14R*)]-13-[(2,6-Dideoxy-3-C-methyl-3-O-methyl-α-L-ribo-hexopyranosyl)oxy]-2,6-diethyl-3,4,10-trihydroxy-3,5,8,10,12,14-hexamethyl-11-[[3,4,6-trideoxy-3-(dimethylamino)-β-D-xylo-hexopyranosyl]oxy]-1-oxa-6-azacyclopentadecan-15-one, Azithromycin N-ethyl
IUPAC Name (2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-11-[(2S,6R)-4-(dimethylamino)-3-hydroxy-6-methyl-tetrahydropyran-2-yl]oxy-2,6-diethyl-3,4,10-trihydroxy-13-[(2R,5S)-5-hydroxy-4-methoxy-4,6-dimethyl-tetrahydropyran-2-yl]oxy-3,5,8,10,12,14-hexamethyl-1-oxa-6-azacyclopentadecan-15-one
SMILES CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2CC(C)(OC)[C@@H](O)C(C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)CC(C3O)N(C)C)[C@](C)(O)C[C@@H](C)CN(CC)[C@H](C)[C@@H](O)[C@]1(C)O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C39H74N2O12/c1-15-28-39(11,47)32(43)25(7)41(16-2)20-21(3)18-37(9,46)34(53-36-30(42)27(40(12)13)17-22(4)49-36)23(5)31(24(6)35(45)51-28)52-29-19-38(10,48-14)33(44)26(8)50-29/h21-34,36,42-44,46-47H,15-20H2,1-14H3/t21-,22-,23+,24-,25-,26?,27?,28-,29+,30?,31+,32-,33+,34-,36+,37-,38?,39-/m1/s1
Formula Weight 762.5242
Percent Purity >95%
Chemical Name or Material Azithromycin N-Ethyl
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.