missing translation for 'onlineSavingsMsg'
Learn More

Azithromycin dihydrate 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer

Product Code. 30325721
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Product Code. Quantity unitSize
30325721 1 mL 1mL
1 options
This item is not returnable. View return policy

Product Code. 30325721

Brand: Dr. Ehrenstorfer DREA10386000AL100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Azithromycin dihydrate
CAS 117772-70-0
Analyte or Component Names Azithromycin dihydrate
Shipping Condition Room Temperature
Molecular Formula C38 H72 N2 O12 . 2 H2 O
Concentration 100 μg/mL
Certifications/Compliance ISO 17034
Synonym Azithromycin dihydrate, (2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-13-[(2,6-Dideoxy-3-C-methyl-3-O-methyl-α-l-ribo-hexopyranosyl)oxy]-2-ethyl-3,4,10-trihydroxy-3,5,6,8,10,12,14-heptamethyl-11-[[3,4,6-trideoxy-3-(dimethylamino)-β-d-xylo-hexopyranosyl]oxy]-1-oxa-6-azacyclopentadecan-15-one dihydrate
InChI Formula InChI=1S/C38H72N2O12.2H2O/c1-15-27-38(10,46)31(42)24(6)40(13)19-20(2)17-36(8,45)33(52-35-29(41)26(39(11)12)16-21(3)48-35)22(4)30(23(5)34(44)50-27)51-28-18-37(9,47-14)32(43)25(7)49-28;;/h20-33,35,41-43,45-46H,15-19H2,1-14H3;2*1H2/t20-,21-,22+,23-,24-,25+,26+,27-,28+,29-,30+,31-,32+,33-,35+,36-,37-,38-;;/m1../s1
SMILES O.O.CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]3O[C@H](C)C[C@@H]([C@H]3O)N(C)C)[C@](C)(O)C[C@@H](C)CN(C)[C@H](C)[C@@H](O)[C@]1(C)O
IUPAC Name (2R,3S,4R,5R,8R,10R,11R,12S,13S,14R)-11-[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy-2-ethyl-3,4,10-trihydroxy-13-[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy-3,5,6,8,10,12,14-heptamethyl-1-oxa-6-azacyclopentadecan-15-one;dihydrate
Solvent or Matrix Acetonitrile
Molecular Weight (g/mol) 785.02
Quantity 1 mL
Formula Weight 784.5297 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.