missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melford Azithromcyin
Description
Product usually ships within 24 hours. Research or further manufacturing use only, not for food or drug use.
Azithromycin is an antibiotic that is commonly used for the following infections caused by certain bacteria.
Specifications
Specifications
| Chemical Name or Material | Azithromcyin |
| CAS | 83905-01-5 |
| Quantity | 1 g |
| Molecular Formula | C38H72N2O12 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?