Learn More
Invitrogen™ AZIN1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5100122
Product Code. 16676604
  • £449.00 / 100µL
Description
Antibody detects endogenous levels of total Antizyme inhibitor 1.
Antizyme inhibitor (AZIN1) protein that positively regulates ornithine decarboxylase (ODC) activity and polyamine uptake. AZI is an enzymatically inactive ODC homolog that counteracts the negative effect of ODC antizymes (AZs) OAZ1, OAZ2 and OAZ3 on ODC activity by competing with ODC for antizyme-binding. Inhibits antizyme-dependent ODC degradation and releases ODC monomers from their inactive complex with antizymes, leading to formation of the catalytically active ODC homodimer and restoring polyamine production.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AZIN1
Polyclonal
Unconjugated
AZIN1
1700085L02Rik; antizyme inhibitor 1; AZI; AZIA1; AZIN1; O OAZIN; OAZI; Oazin; ODC antizyme inhibitor; ODC1L; Ornithine decarboxylase antizyme inhibitor
Rabbit
Affinity chromatography
RUO
51582, 54375, 58961
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
O14977, O35484, Q63764
AZIN1
A synthesized peptide derived from human AZIN1(Accession O14977), corresponding to amino acid residues A325-V375.
100 μL
Primary
Human, Mouse, Rat