missing translation for 'onlineSavingsMsg'
Learn More

Azilsartan medoxomil, MedChemExpress

Product Code. 16488747
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16488747

Brand: Medchem Express HY14736

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Azilsartan medoxomil(TAK 491) is an orally administered angiotensin II receptor type 1 antagonist with IC50 of 0.62 nM, which used in the treatment of adults with essential hypertension.

Azilsartan medoxomil(TAK 491) is an orally administered angiotensin II receptor type 1 antagonist with IC50 of 0.62 nM, which used in the treatment of adults with essential hypertension.

Specifications

Chemical Name or Material Azilsartan medoxomil
CAS 863031-21-4
Color White
Purity Grade Notes Research
Molecular Formula C30H24N4O8
Quantity 5 mg
Synonym TAK-491
Solubility Information DMSO : 125 mg/mL (219.87 mM; Need ultrasonic)
SMILES O=C(C1=C2C(N=C(OCC)N2CC3=CC=C(C4=CC=CC=C4C5=NC(ON5)=O)C=C3)=CC=C1)OCC6=C(C)OC(O6)=O
Molecular Weight (g/mol) 568.53
Formula Weight 568.53
Percent Purity 99.88%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.