missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Azidothymidine

Product Code. 18767958
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18767958 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18767958 Supplier R&D Systems™ Supplier No. 4150/50

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Selective reverse transcriptase inhibitor; antiretroviral

Selective reverse transcriptase inhibitor. Exhibits 100-fold selectivity for HIV reverse transcriptase over DNA polymerase α. Suppresses HIV-1 replication and enhances cell viability in HIV-1 infected T cells. Attenuates growth of multiple myeloma (MM) cells in vitro and reduces growth of MM tumor xenografts in mice. Increases differentiation iPSCs into cortical neurons and reduces the over-proliferation of neural precursors. Orally bioavailable and brain penetrant. Antiretroviral agent. Also decreases CRISPR-mediated homology directed repair (HDR) efficiency.
TRUSTED_SUSTAINABILITY

Specifications

Purity 99%
Molecular Formula 3 '-Azido-3 '-deoxythymidine
CAS 30516-87-1
Content And Storage -20°C
Quantity 50 mg
Product Type Azidothymidine
Molecular Weight (g/mol) M.W. (Observed): 267.24
Product Line Tocris
Inhibitors Azidothymidine

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.