missing translation for 'onlineSavingsMsg'
Learn More

Azido-PEG3-maleimide Kit

Product Code. 15467169
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Product Code. Quantity unitSize
15467169 100 mg 100mg
15457169 25 mg 25mg
2 options
This item is not returnable. View return policy

Product Code. 15467169

Brand: Thermo Scientific Alfa Aesar J65984.MC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

C15H23N5O6

An azido containing reagent with PEG spacer and sulfhydryl reactivity. The compound is not stable upon prolonged storage and is sold as a kit.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An azido containing reagent with PEG spacer and sulfhydryl reactivity. The compound is not stable upon prolonged storage and is sold as a kit.

Solubility
Difficult to mix.

Notes
Kit contains two components: a) 1.1 eq. of azido-PEG3-amine as an oil; b) 1.0 eq. of maleimide-NHS ester as a solid.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C15H23N5O6
MDL Number MFCD22380764
Synonym N-(11-Azido-3,6,9-trioxaundecyl)-3-(N-maleimidyl)propionamide

Specifications

Quantity 100 mg
UN Number UN2735
Solubility Information Difficult to mix.
Formula Weight 369.37
Chemical Name or Material Azido-PEG3-maleimide Kit
What storage conditions should I use for the Azido-PEG3-maleimide Kit?

The Azido-PEG3-maleimide Kit should be stored in the dark at -20°C.

What is the protocol for Azide-PEG3-maleimide generation with the Azido-PEG3-maleimide Kit?

We recommend the following protocol for Azide-PEG3-maleimide generation with the Azido-PEG3-maleimide Kit:

  1. For the 25 mg kit, add 1 mL of a dry water-miscible solvent (like DMF or DMSO) to the azido-PEG-amine and shake for approximately 30 seconds to ensure mixing. For the 100 mg kit add 2.5 mL of solvent.
  2. Place the maleimide-NHS ester under nitrogen or dry atmosphere and slowly add the solution from step 1 with stirring or shaking.
  3. Stir or shake the reaction mixture for 30 minutes at room temperature. The progress of the reaction can be followed by Thin Layer Chromatograph.
    Note: For TLC, the solvent is typically methanol:methylene chloride (1:20), run on a silica gel normal phase plate and developed with a potassium permanganate spray. The Rf of the azide-PEG3-maleimide is slightly lower than the Rf for the maleimide-NHS ester. When the reaction is complete, it will be one clean spot on the plate.
  4. The reaction product will be ready-to-use azide-PEG3-maleimide.
The azide-PEG3-maleimide is present at ~ 75 mM if 1 mL of solvent is used in step 1. The 25 mg kit produces 0.075 mmol of azide-PEG3-maleimide.

compliance-icons
Product Identifier
  • 1-Amino-11-azido-3,6,9-trioxaundecane
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : 1-Amino-11-azido-3,6,9-trioxaundecane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.