missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azido-PEG11-biotin conjugate, Thermo Scientific™
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
An azide-biotin conjugate with a long PEG spacer, Biotinylation reagent for labeling alkyne containing molecules or biomolecules. The azide group reacts with alkynes to form a stable triazole linkage facilitating the introduction of biotin into your alkyne modified system of interest.
Solubility
Soluble in DMSO, DMF and water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
An azide-biotin conjugate with a long PEG spacer, Biotinylation reagent for labeling alkyne containing molecules or biomolecules. The azide group reacts with alkynes to form a stable triazole linkage facilitating the introduction of biotin into your alkyne modified system of interest.
Solubility
Soluble in DMSO, DMF and water.
Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications
Specifications
| Color | White to Gray |
| Quantity | 100 mg |
| Formula Weight | 796.96 |
| Physical Form | Solid |
| Chemical Name or Material | Azido-PEG11-biotin conjugate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction