missing translation for 'onlineSavingsMsg'
Learn More

Azido-PEG11-biotin conjugate, Thermo Scientific™

Product Code. 15416759
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 15416759

Brand: Thermo Scientific Alfa Aesar J65147.MB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

An azide-biotin conjugate with a long PEG spacer

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
An azide-biotin conjugate with a long PEG spacer, Biotinylation reagent for labeling alkyne containing molecules or biomolecules. The azide group reacts with alkynes to form a stable triazole linkage facilitating the introduction of biotin into your alkyne modified system of interest.

Solubility
Soluble in DMSO, DMF and water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C34H64N6O13S
MDL Number MFCD21363313
Synonym 35-[(R)-(+)-Biotinylamino]-1-azido-3,6,9,12,15,18,21,24,27,30,33-undecaoxapentatriacontane

Specifications

Color White to Gray
Quantity 25 mg
Formula Weight 796.96
Physical Form Solid
Chemical Name or Material Azido-PEG11-biotin conjugate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.