missing translation for 'onlineSavingsMsg'
Learn More

Azido-erythro-sphingosine, TRC

Product Code. 30179605
Change view
Click to view available options
Quantity:
1 mg
25 mg
250 mg
Unit Size:
1mg
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30179605 25 mg 25mg
30179519 1 mg 1mg
30179193 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30179605 Supplier Toronto Research Chemicals Supplier No. TRCA92650025MG
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103348-49-8
Molecular Formula C18 H35 N3 O2
Molecular Weight (g/mol) 325.49
IUPAC Name (E,2S,3R)-2-azidooctadec-4-ene-1,3-diol
SMILES CCCCCCCCCCCCC\C=C\[C@@H](O)[C@H](CO)N=[N+]=[N-]

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C18H35N3O2/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-18(23)17(16-22)20-21-19/h14-15,17-18,22-23H,2-13,16H2,1H3/b15-14+/t17-,18+/m0/s1
Formula Weight 325.2729
Chemical Name or Material Azido-erythro-sphingosine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.