Learn More
Invitrogen™ AZI2 Recombinant Rabbit Monoclonal Antibody (24GB3555)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA556948
 View more versions of this product
Product Code. 30276473
  • £126.00 / 20µL
Description
AZI2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
AZI2, or NAP1, contributes to the activation of NFKB (Fujita et al., 2003 ).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
AZI2
Recombinant Monoclonal
Unconjugated
AZI2
5-azacytidine induced 2; 5-azacytidine induced gene 2; 5-azacytidine-induced protein 2; AA410145; Az2; AZI2; Nak-associated protein 1; NAP1; NF-kappa-B-activating kinase-associated protein 1; TBKBP2; Tbkp2; TILP
Rabbit
Affinity Chromatography
RUO
27215, 316051, 64343
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB3555
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q4KMA0, Q9H6S1, Q9QYP6
AZI2
A synthesized peptide derived from human AZI2 (aa 280-392).
20 μL
Primary
Human, Mouse, Rat