missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AZI1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101378
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total AZI1.
AZI1, also named as CEP131 and KIAA1118, may play a role in spermatogenesis. It is located at the ciliary base and required for cilium formation.
Specifications
| AZI1 | |
| Polyclonal | |
| Unconjugated | |
| CEP131 | |
| 5-azacytidine induced 1; 5-azacytidine induced gene 1; 5-azacytidine-induced protein 1; Az1; Azi; AZI1; centrosomal protein 131; centrosomal protein 131 kDa; centrosomal protein 131kDa; centrosomal protein of 131 kDa; CEP131; KIAA1118; Pre-acrosome localization protein 1; ZA1 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 12009, 22994 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q62036, Q9UPN4 | |
| CEP131 | |
| A synthesized peptide derived from human AZI1, corresponding to a region within C-terminal amino acids. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction