missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ AZGP1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA580366
This item is not returnable.
View return policy
Description
This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human Zinc Alpha 2 Glycoprotein.
Alpha-2-glycoprotein 1, zinc-binding (AZGP1) is a gene encoding a protein that belongs to the immunoglobulin superfamily. This protein is abundant in various body fluids, such as blood plasma and milk, and is involved in lipid mobilization. AZGP1 is expressed in numerous tissues, including the liver, prostate, and adipose tissue, where it plays a significant role in the regulation of lipid metabolism and adipocyte function. It acts as an adipokine, influencing the breakdown and mobilization of fats, thereby contributing to energy homeostasis. In cancer research, AZGP1 has been highlighted for its potential as a prognostic biomarker, particularly in prostate and breast cancer, where lower expression levels are often associated with poor prognosis. Recent studies have also linked AZGP1 to cachexia, an extreme form of weight loss and muscle wasting, by promoting lipid mobilization in adipose tissue and influencing systemic catabolic states. Due to its multifaceted roles in lipid metabolism and disease processes, AZGP1 is considered a promising target for therapeutic interventions in metabolic and cancer-related disorders.
Specifications
| AZGP1 | |
| Polyclonal | |
| Unconjugated | |
| AZGP1 | |
| alpha-2 - glycoprotein 1 zinc; alpha-2 - glycoprotein 1, zinc; alpha-2-glycoprotein 1, zinc; alpha-2-glycoprotein 1, zinc-binding; Alpha-2-glycoprotein, zinc; Azgp1; major histocompatibility complex class I-like protein; testicular tissue protein Li 227; ZA2G; ZA2GA; Zag; Zinc a 2 glycoprotein; Zinc a2glycoprotein; Zinc alpha2glycoprotein; zinc alpha-2-glycoprotein 1; Zinc a 2 glycoprotein; Zinc a2glycoprotein; zinc-alpha-2-glycoprotein; Zn - alpha2 - glycoprotein; Zn a 2 glycoprotein; Zn a 2 GP; Zn alpha 2 glycoprotein; Zn alpha 2 GP; Zn a 2 glycoprotein; Zn a 2 GP; Zna2gp; zn-alpha-2-glycoprotein; Zn-alpha2-glycoprotein; zn-alpha-2-GP; ZNGP1 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 563 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 1 mg/mL | |
| PBS with no preservative | |
| P25311 | |
| AZGP1 | |
| Recombinant Human Zinc Alpha 2 Glycoprotein Protein (Met1-Ser298). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction