missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AZGP1, Mouse anti-Human, Clone: Zagy-1, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova MAB16118.100ug
This item is not returnable.
View return policy
Specifications
| AZGP1 | |
| Monoclonal | |
| Unconjugated | |
| In PBS (10% glycerol, 0.02% sodium azide). | |
| ZA2G/ZAG | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry | |
| Zagy-1 | |
| Flow Cytometry (1:100) The optimal working dilution should be determined by the end user. | |
| alpha-2-glycoprotein 1, zinc-binding | |
| AZGP1 | |
| Recombinant protein corresponding to human AZGP1. | |
| RUO | |
| 563 | |
| For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction