missing translation for 'onlineSavingsMsg'
Learn More

Azetidine-3-carboxylic acid, 98+%

Product Code. 15468917
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15468917 1 g 1g
15478917 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15468917 Supplier Thermo Scientific Alfa Aesar Supplier No. H52294.03
 Restricted Item
Estimated Shipment: 14-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 36476-78-5 | C4H7NO2 | 101.11 g/mol

Azetidine-3-carboxylic acid derivatives has potential biological and foldameric applications. Azetidine-3-carboxylic acid, a fairly strained molecule provides oxadiazoles in good yields. Azetidine-3-carboxylic acid is used as a chemical hybridising agent

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Azetidine-3-carboxylic acid derivatives has potential biological and foldameric applications. Azetidine-3-carboxylic acid, a fairly strained molecule provides oxadiazoles in good yields. Azetidine-3-carboxylic acid is used as a chemical hybridising agent

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 36476-78-5
Molecular Formula C4H7NO2
Molecular Weight (g/mol) 101.11
MDL Number MFCD00191763
InChI Key GFZWHAAOIVMHOI-UHFFFAOYSA-N
Synonym 3-azetidinecarboxylic acid, 3-azetanecarboxylic acid, 3-carboxyazetidine, caswell no. 063c, h-aze 3-oh, azetane-3-carboxylic acid, epa pesticide chemical code 128830, 3-azetidine carboxylic acid, l-azetidine-3-carboxylic acid, azetidine-3-carboxylicacid
PubChem CID 93192
IUPAC Name azetidine-3-carboxylic acid
SMILES OC(=O)C1CNC1

Specifications

Melting Point ∽250°C (decomposition)
Quantity 1 g
Solubility Information Soluble in water.
Formula Weight 101.11
Percent Purity ≥98%
Chemical Name or Material Azetidine-3-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.