missing translation for 'onlineSavingsMsg'
Learn More

Azemiglitazone, MedChemExpress

Product Code. 16494168
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494168 10 mg 10mg
16480517 50 mg 50mg
16456877 5 mg 5mg
16415136 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16494168 Supplier Medchem Express Supplier No. HY108022

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Azemiglitazone (MSDC-0602), a PPARγ-sparing thiazolidinedione (TZD), interacts with the mitochondrial pyruvate carrier (MPC) and inhibits its activity and has the potential for type 2 diabetes study with reducing risk of PPARγ-mediated side effects.

Azemiglitazone (MSDC-0602), a PPARγ-sparing thiazolidinedione (TZD), interacts with the mitochondrial pyruvate carrier (MPC) and inhibits its activity and has the potential for type 2 diabetes study with reducing risk of PPARγ-mediated side effects.

Specifications

Chemical Name or Material Azemiglitazone
CAS 1133819-87-0
Color White
Purity Grade Notes Research
Molecular Formula C19H17NO5S
Quantity 10 mg
Synonym MSDC-0602
Solubility Information DMSO : 125 mg/mL (336.56 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1C(SC(N1)=O)CC2=CC=C(OCC(C3=CC(OC)=CC=C3)=O)C=C2
Molecular Weight (g/mol) 371.41
Formula Weight 371.41
Percent Purity 97.24%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.