missing translation for 'onlineSavingsMsg'
Learn More

Azelastine N-Oxide (Mixture of Diastereomers), TRC

Product Code. 30040432
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30040432 50 mg 50mg
30041387 10 mg 10mg
30041071 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30040432 Supplier Toronto Research Chemicals Supplier No. TRCA80826050MG
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 640279-88-5
Molecular Formula C22 H24 Cl N3 O2
Molecular Weight (g/mol) 397.9
Synonym 1(2H)-Phthalazinone, 4-[(4-chlorophenyl)methyl]-2-(hexahydro-1-methyl-1-oxido-1H-azepin-4-yl)-, Azelastine N-Oxide
IUPAC Name 4-[(4-chlorophenyl)methyl]-2-(1-methyl-1-oxidoazepan-1-ium-4-yl)phthalazin-1-one
SMILES C[N+]1([O-])CCCC(CC1)N2N=C(Cc3ccc(Cl)cc3)c4ccccc4C2=O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C22H24ClN3O2/c1-26(28)13-4-5-18(12-14-26)25-22(27)20-7-3-2-6-19(20)21(24-25)15-16-8-10-17(23)11-9-16/h2-3,6-11,18H,4-5,12-15H2,1H3
Formula Weight 397.1557
Percent Purity >95%
Chemical Name or Material Azelastine N-Oxide (Mixture of Diastereomers)
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.