missing translation for 'onlineSavingsMsg'
Learn More

AZD9496 maleate, MedChemExpress

Product Code. 16438137
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16438137

Brand: Medchem Express HY12870A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AZD9496 maleate is a potent and selective estrogen receptor (ERα) antagonist with IC50 of 0.28 nM. AZD9496 maleate is an orally bioavailable selective oestrogen receptor degrader (SERD).

AZD9496 maleate is a potent and selective estrogen receptor (ERα) antagonist with IC50 of 0.28 nM. AZD9496 maleate is an orally bioavailable selective oestrogen receptor degrader (SERD).

Specifications

Chemical Name or Material AZD9496 maleate
CAS 1639042-28-6
Color Brown
Purity Grade Notes Research
Molecular Formula C29H29F3N2O6
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (179.04 mM)
SMILES O=C(O)/C=C/C1=CC(F)=C([C@H]2N(CC(C)(F)C)[C@H](C)CC3=C2NC4=C3C=CC=C4)C(F)=C1.O=C(O)/C=C\C(O)=O
Molecular Weight (g/mol) 558.55
Formula Weight 558.55
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.