missing translation for 'onlineSavingsMsg'
Learn More

AZD8797, MedChemExpress

Product Code. 16423396
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16423396 25 mg 25mg
16488527 5 mg 5mg
16456346 100 mg 100mg
16455888 10 mg 10mg
16452037 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16423396 Supplier Medchem Express Supplier No. HY13848

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AZD8797 (KAND567) is an allosteric non-competitive and orally active antagonist of the human CX3CR1 receptor; antagonizes CX3CR1 and CXCR2 with Kis of 3.9 and 2800 nM, respectively.

AZD8797 (KAND567) is an allosteric non-competitive and orally active antagonist of the human CX3CR1 receptor

Specifications

Chemical Name or Material AZD8797
Physical Form Solid
CAS 911715-90-7
Color Light Yellow
Quantity 25 mg
Purity Grade Notes Research
Molecular Formula C19H25N5OS2
For Use With (Application) COVID-19-immunoregulation
Synonym KAND567
Solubility Information DMSO : ≥ 150 mg/mL (371.69 mM)
SMILES CC(C)C[C@@H](NC1=NC(S[C@@H](C)C2=CC=CC=C2)=NC3=C1SC(N)=N3)CO
Molecular Weight (g/mol) 403.56
Formula Weight 403.56
Percent Purity 98.65%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.