missing translation for 'onlineSavingsMsg'
Learn More

AZD7687, MedChemExpress

Product Code. 16459147
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16459147 5 mg 5mg
16452667 50 mg 50mg
16446578 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16459147 Supplier Medchem Express Supplier No. HY15497
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress AZD7687 is a potent, selective, reversible and orally active diacylglycerol acyltransferase 1 (DGAT1) inhibitor with an IC50 of 80 nM for human DGAT1. AZD7687 can be used for type 2 diabetes mellitus and obesity research.

AZD7687 is a potent, selective, reversible and orally active diacylglycerol acyltransferase 1 (DGAT1) inhibitor with an IC50 of 80 nM for human DGAT1. AZD7687 can be used for type 2 diabetes mellitus and obesity research.

Specifications

Chemical Name or Material AZD7687
CAS 1166827-44-6
Color Pink
Purity Grade Notes Research
Molecular Formula C21H25N3O3
Quantity 5 mg
Solubility Information DMSO : ≥ 50 mg/mL (136.08 mM)
SMILES CC1=C(C(N)=O)N=C(C2=CC=C([C@H]3CC[C@H](CC(O)=O)CC3)C=C2)C(C)=N1
Molecular Weight (g/mol) 367.44
Formula Weight 367.44
Percent Purity 99.04%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.