missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AZD5582, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Description
AZD5582 is an antagonist of the inhibitor of apoptosis proteins (IAPs), which binds to the BIR3 domains cIAP1, cIAP2, and XIAP with IC50s of 15, 21, and 15 nM, respectively. AZD5582 induces apoptosis.
Specifications
Specifications
| Chemical Name or Material | AZD5582 |
| CAS | 1258392-53-8 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C58H78N8O8 |
| Quantity | 5 mg |
| Solubility Information | DMSO : ≥ 66.25 mg/mL (65.25 mM) |
| SMILES | CN[C@@H](C)C(N[C@H](C(N1[C@H](C(N[C@H]2C3=CC=CC=C3C[C@H]2OCC#CC#CCO[C@H]4[C@@H](NC([C@@H]5CCCN5C([C@@H](NC([C@H](C)NC)=O)C6CCCCC6)=O)=O)C(C=CC=C7)=C7C4)=O)CCC1)=O)C8CCCCC8)=O |
| Molecular Weight (g/mol) | 1015.29 |
| Formula Weight | 1015.29 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction