missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AZD5582, MedChemExpress
Description
AZD5582 is an antagonist of the inhibitor of apoptosis proteins (IAPs), which binds to the BIR3 domains cIAP1, cIAP2, and XIAP with IC50s of 15, 21, and 15 nM, respectively. AZD5582 induces apoptosis.
Specifications
Specifications
| Chemical Name or Material | AZD5582 |
| CAS | 1258392-53-8 |
| Color | Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C58H78N8O8 |
| Quantity | 5 mg |
| Solubility Information | DMSO : ≥ 66.25 mg/mL (65.25 mM) |
| SMILES | CN[C@@H](C)C(N[C@H](C(N1[C@H](C(N[C@H]2C3=CC=CC=C3C[C@H]2OCC#CC#CCO[C@H]4[C@@H](NC([C@@H]5CCCN5C([C@@H](NC([C@H](C)NC)=O)C6CCCCC6)=O)=O)C(C=CC=C7)=C7C4)=O)CCC1)=O)C8CCCCC8)=O |
| Molecular Weight (g/mol) | 1015.29 |
| Formula Weight | 1015.29 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?