missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AZD5363, TRC
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
Specifications
Specifications
| Chemical Name or Material | AZD5363 |
| CAS | 1143532-39-1 |
| Molecular Formula | C21 H25 Cl N6 O2 |
| Quantity | 1 mg |
| Synonym | 4-Piperidinecarboxamide, 4-amino-N-[(1S)-1-(4-chlorophenyl)-3-hydroxypropyl]-1-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)- (ACI), 4-Amino-N-[(1S)-1-(4-chlorophenyl)-3-hydroxypropyl]-1-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)-4-piperidinecarboxamide (ACI), (S)-4-Amino-N-(1-(4-chlorophenyl)-3-hydroxypropyl)-1-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)piperidine-4-carboxamide, 4-amino-N-[(1S)-1-(4-chlorophenyl)-3-hydroxypropyl]-1-(1H-pyrrolo[2,3-d]pyrimidin-4-yl)piperidine-4-carboxamide, AZD 5363, AZD-5363, AZD5363, Capivasertib |
| InChI Formula | InChI=1S/C21H25ClN6O2/c22-15-3-1-14(2-4-15)17(6-12-29)27-20(30)21(23)7-10-28(11-8-21)19-16-5-9-24-18(16)25-13-26-19/h1-5,9,13,17,29H,6-8,10-12,23H2,(H,27,30)(H,24,25,26)/t17-/m0/s1 |
| SMILES | NC1(CCN(CC1)c2ncnc3[nH]ccc23)C(=O)N[C@@H](CCO)c4ccc(Cl)cc4 |
| IUPAC Name | 4-amino-N-[(1S)-1-(4-chlorophenyl)-3-hydroxypropyl]-1-(7H-pyrrolo[2,3-d]pyrimidin-4-yl)piperidine-4-carboxamide |
| Molecular Weight (g/mol) | 428.92 |
| Formula Weight | 428.1728 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction