missing translation for 'onlineSavingsMsg'
Learn More

AZD4547, MedChemExpress

Product Code. 16495648
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16495648 10 mg 10mg
16486166 100 mg 100mg
16481817 50 mg 50mg
16461076 200 mg 200mg
16448347 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16495648 Supplier Medchem Express Supplier No. HY13330
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress AZD4547 is a potent inhibitor of the FGFR family with IC50s of 0.2 nM, 2.5 nM, 1.8 nM, and 165 nM for FGFR1, FGFR2, FGFR3, and FGFR4, respectively.

AZD4547 is a potent inhibitor of the FGFR family with IC50s of 0.2 nM, 2.5 nM, 1.8 nM, and 165 nM for FGFR1, FGFR2, FGFR3, and FGFR4, respectively.

Specifications

Chemical Name or Material AZD4547
CAS 1035270-39-3
Color White
Purity Grade Notes Research
Molecular Formula C26H33N5O3
Quantity 10 mg
Solubility Information DMSO : 125 mg/mL (269.65 mM; Need ultrasonic)
SMILES C[C@H](C1)N[C@@H](C)CN1C2=CC=C(C(NC3=NNC(CCC4=CC(OC)=CC(OC)=C4)=C3)=O)C=C2
Molecular Weight (g/mol) 463.57
Formula Weight 463.57
Percent Purity 98.87%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.